Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-20R alpha Antibody (038), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289469AF750
Description
IL-20R alpha Monoclonal specifically detects IL-20R alpha in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IL-20R alpha | |
Monoclonal | |
Alexa Fluor 750 | |
class II cytokine receptor ZCYTOR7, CRF2-8, Cytokine receptor class-II member 8, Cytokine receptor family 2 member 8, FLJ40993, IL-20R1IL-20 receptor subunit alpha, IL-20RA, IL-20R-alpha, interleukin 20 receptor, alpha, interleukin-20 receptor I, interleukin-20 receptor subunit alpha, ZcytoR7 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-20R alpha (NP_055247.3; Met 1-Lys 250). | |
0.1 mL | |
Primary | |
No cross-reactivity in ELISA with: Human IFNgR1 Human IL10RB | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
38 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
53832 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction