Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-20R alpha Antibody (038), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289469PCP
Description
IL-20R alpha Monoclonal antibody specifically detects IL-20R alpha in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
IL-20R alpha | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
53832 | |
Store at 4C in the dark. | |
IgG |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
038 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
class II cytokine receptor ZCYTOR7, CRF2-8, Cytokine receptor class-II member 8, Cytokine receptor family 2 member 8, FLJ40993, IL-20R1IL-20 receptor subunit alpha, IL-20RA, IL-20R-alpha, interleukin 20 receptor, alpha, interleukin-20 receptor I, interleukin-20 receptor subunit alpha, ZcytoR7 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-20R alpha (Accession#: NP_055247.3; Met 1-Lys 250). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction