Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-20R alpha Antibody, Novus Biologicals™
SDP

Catalog No. p-5066657 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056409T 0.025 mg
NB10056409 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056409T Supplier Novus Biologicals Supplier No. NB10056409SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL-20R alpha Polyclonal specifically detects IL-20R alpha in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen IL-20R alpha
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Flow Cytometry 2-5 ug/10^6 cells
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Alias class II cytokine receptor ZCYTOR7, CRF2-8, Cytokine receptor class-II member 8, Cytokine receptor family 2 member 8, FLJ40993, IL-20R1IL-20 receptor subunit alpha, IL-20RA, IL-20R-alpha, interleukin 20 receptor, alpha, interleukin-20 receptor I, interleukin-20 receptor subunit alpha, ZcytoR7
Gene Symbols IL20RA
Host Species Rabbit
Immunogen Amino acids 159-175 of human IL-20 receptor alpha protein were used as the immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53832
Test Specificity Both the expect ∼70 kD band and an unknown ∼90 kD band are seen in A375 cell lysate and are blocked by the immunizing peptide. The ∼90 kD band alone is seen in A431 cell lysate, and it is also blocked by the immunizing peptide. This antibody was made against a portion of the extracellular domain of IL-20RA. Permeabilization of the cells during testing may affect antibody binding.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.