Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-21 Antibody (14k5H3), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227336R
Description
IL-21 Monoclonal specifically detects IL-21 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
IL-21 | |
Monoclonal | |
DyLight 550 | |
Q9HBE4 | |
IL21 | |
Full length recombinant protein | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity, Protein Kinase, Signal Transduction | |
59067 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
14k5H3 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction