Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-21 Antibody (227), DyLight 755, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326073IR
Description
IL-21 Monoclonal antibody specifically detects IL-21 in Mouse samples. It is validated for ELISASpecifications
IL-21 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
227 | |
ELISA | |
IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform | |
Rabbit immunized with purified, recombinant Mouse IL-21 (Uniprot#: Q9ES17, Pro25-Ser146) | |
0.1 mL | |
Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Protein Kinase, Signal Transduction | |
59067 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction