Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-21 Antibody (RM0268-6G53), Alexa Fluor™ 750, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP211696AF750
Description
IL-21 Monoclonal specifically detects IL-21 in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
IL-21 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
IL21 | |
Mouse recombinant IL-21 | |
0.1 mL | |
Primary | |
Mouse | |
IgG2 |
Western Blot, Flow Cytometry | |
RM0268-6G53 | |
Western Blot, Flow Cytometry | |
IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform | |
Rat | |
Protein A or G purified | |
Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Protein Kinase, Signal Transduction | |
59067.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction