Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-21 Antibody (RM0268-6G53), DyLight 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP211696V
Description
IL-21 Monoclonal specifically detects IL-21 in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
IL-21 | |
Monoclonal | |
DyLight 405 | |
IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Flow Cytometry | |
RM0268-6G53 | |
Western Blot, Flow Cytometry | |
IL21 | |
Mouse recombinant IL-21 | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity, Protein Kinase, Signal Transduction | |
59067 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction