Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-21 Rabbit anti-Human, Clone: 145, Novus Biologicals™

Rabbit Monoclonal Antibody
$524.00
Specifications
Antigen | IL-21 |
---|---|
Clone | 145 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
IL-21 Monoclonal specifically detects IL-21 in Human samples. It is validated for ELISA.Specifications
IL-21 | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform | |
Recombinant Human IL-21 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
145 | |
ELISA | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Protein Kinase, Signal Transduction | |
0.2 um filtered solution in PBS | |
59067 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title