Learn More
IL-21 Monoclonal Antibody (FFA21), eFluor™ 450, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$200.00 - $454.00
Specifications
Antigen | IL-21 |
---|---|
Clone | FFA21 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
48-721-180
![]() |
Invitrogen™
48721180 |
25 μg |
Each for $200.00
|
![]() |
|||||
48-721-182
![]() |
Invitrogen™
48721182 |
100 μg |
Each for $454.00
|
![]() |
|||||
Description
Description: This FFA21 monoclonal antibody reacts with mouse interleukin-21 (IL-21). IL-21 is a 17 kDa immunomodulatory cytokine produced mainly by NKT, T helper (Th) 17 and T follicular helper (TFH) cells. In TFH cells, IL-21 expression leads to autocrine signaling through the IL-21 receptor (IL-21R) and STAT3, which leads to additional transcriptional activation by Bcl6. As with IFN gamma for Th1, IL-4 for Th2 and IL-17A for Th17, IL-21 is critical for TFH cell effector function. This cytokine plays a role in T cell-dependent B cell differentiation into plasma cells and memory cells, stimulation of IgG production and induction of apoptotic signaling in naive B cells. In Th17 cells, IL-21 expression and autocrine feedback through STAT3, IRF4 and ROR gamma t lead to upregulation of the IL-23R, thereby preparing Th17 cells for maturation and maintece by the inflammatory cytokine IL-23. While upregulating IRF4 and ROR gamma t, IL-21 also mediates the downregulation of Foxp3. High levels of IL-21 are present in chemically-induced colitis models. IL-21-deficient mice are protected from developing colitis upon chemical treatment by their inability to upregulate Th17-associated molecules. Preliminary data suggest that clone FFA21 recognizes a different epitope than anti-mouse IL-21 clone mhalx21 (cat. 51-7213). Applications Reported: This FFA21 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.
IL-21 is a 17 kDa immunomodulatory cytokine produced mainly by Natural Killer Cells (NKT), T helper (Th) 17 and T follicular helper (TFH) cells. In TFH cells, IL-21 expression leads to autocrine signaling through the IL-21 receptor (IL-21R) and STAT3, which leads to additional transcriptional activation by Bcl6. As with IFN gamma for Th1 and IL-4 for Th2 cells, IL-21 is critical for TFH cell development and effector function. IL-21 plays a role in T cell-dependent B cell differentiation into plasma cells and memory cells, stimulation of IgG production and induction of apoptotic signaling in naive B cells. In Th17 cells, IL-21 expression and autocrine feedback through STAT3, IRF4 and ROR gamma t lead to upregulation of the IL-23R, thereby preparing Th17 cells for maturation and maintenance by the inflammatory cytokine IL-23. While upregulating IRF4 and ROR gamma t, IL-21 also mediates the downregulation of Foxp3. IL-21 deficient mice are protected from developing colitis upon chemical treatment by their inability to upregulate Th17-associated molecules. In comparison, elevated levels of IL-21 are present in chemically-induced colitis mouse models.Specifications
IL-21 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CVID11; CX2; il 21; IL21; IL-21; ILN; Interleukin; interleukin 21; Interleukin21; interleukin-21; interleukin-21 isoform; porcine interleukin-21; Za11 | |
IL21 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
IL21 |
FFA21 | |
Flow Cytometry | |
eFluor 450 | |
Rat | |
Mouse | |
Q9ES17 | |
60505 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.