Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-21R Antibody (033), Alexa Fluor™ 594 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289904AF594
Description
IL-21R Monoclonal antibody specifically detects IL-21R in Human samples. It is validated for ELISASpecifications
IL-21R | |
Monoclonal | |
Alexa Fluor 594 | |
CD360 antigen, IL-21 receptor, IL-21R, interleukin 21 receptor, interleukin-21 receptor, MGC10967, NILR, Novel interleukin receptor | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-21R (Q9HBE5-1; Met 1-Pro 236). | |
0.1 mL | |
Chemokines and Cytokines, Cytokine Research, Immunology | |
50615 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
033 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction