Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-21R Antibody (152512), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB9911E
This item is not returnable.
View return policy
Description
IL-21R Monoclonal antibody specifically detects IL-21R in Human samples. It is validated for Flow CytometrySpecifications
IL-21R | |
Monoclonal | |
DyLight 405 | |
CD360 antigen, IL-21 receptor, IL-21R, interleukin 21 receptor, interleukin-21 receptor, MGC10967, NILR, Novel interleukin receptor | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-21 R, Cys20-Pro236, Accession # Q9HBE5 | |
0.5 mL | |
Chemokines and Cytokines, Cytokine Research, Immunology | |
50615 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry | |
152512 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction