Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-21R Monoclonal Antibody (4A9), Functional Grade, In Vivo GOLD™, Leinco Technologies™
SDP

Catalog No. 502544139
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4139 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4139 Supplier Leinco Technologies Inc Supplier No. I478100MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specificity: Clone 4A9 activity is directed against murine Interleukin-21 Receptor (CD360). Endotoxin Level: <1.0 EU/mg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -70C. Avoid Repeated Freeze Thaw Cycles.

IL-21R Monoclonal antibody specifically detects IL-21R in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunoprecipitation, Inhibition Assays

Specifications

Antigen IL-21R
Applications Flow Cytometry, Functional Assay, Immunoprecipitation, Inhibition Assays
Classification Monoclonal
Clone 4A9
Concentration ≥5.0 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2-7.4
Gene Il21r
Gene Accession No. Q9JHX3
Gene Alias CD360; IL-21 receptor; Il21r; IL-21R; interleukin 21 receptor; interleukin-21 receptor; LR-beta; lymphocyte receptor beta; MGC10967; NILR; novel cytokine receptor 8; Novel interleukin receptor; NR8; UNQ3121/PRO10273
Host Species Rat
Immunogen Rat YB2/0 hybridoma transfectant cell line expressing truncated IL-21R
Purification Method Protein A/G
Quantity 100 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60504
Target Species Mouse
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.