Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-22 Antibody (140301), Alexa Fluor™ 700, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB582N
Description
IL-22 Monoclonal antibody specifically detects IL-22 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
IL-22 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
140301 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL-D110, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
E. coli-derived recombinant mouse IL-22, Leu34-Val179, Accession # Q9JJY9 | |
0.5 mL | |
Biologically Active Proteins, Cytokine Research, Innate Immunity | |
50616 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction