Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-22 Antibody (RM0269-8C61), Alexa Fluor™ 647, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP211699AF647
Description
IL-22 Monoclonal specifically detects IL-22 in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
IL-22 | |
Monoclonal | |
Alexa Fluor 647 | |
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL-D110, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Flow Cytometry | |
RM0269-8C61 | |
Western Blot, Flow Cytometry | |
IL22 | |
Mouse recombinant IL-22 | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research, Innate Immunity | |
50616 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction