Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-22 Mouse anti-Human, Mouse, Alexa Fluor 647, Clone: MH22B2, BD Pharmingen™
Mouse Monoclonal Antibody
$182.00 - $402.00
Specifications
Antigen | IL-22 |
---|---|
Clone | MH22B2 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
IL-22 Monoclonal antibody specifically detects IL-22 in Human, Mouse samples. It is validated for Intracellular staining (Flow Cytometry)Specifications
IL-22 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human IL-22 Recombinant Protein | |
Primary |
MH22B2 | |
Flow Cytometry | |
Alexa Fluor 647 | |
Mouse | |
Human, Mouse | |
interleukin-22; IL-10-related T-cell-derived inducible factor; IL-TIF; ILTIF; IL-22a, ilTIFa; Iltif; TIFa | |
IgG2a κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title