Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-22 Mouse anti-Mouse, Rat, Clone: MT231, Novus Biologicals™

Mouse Monoclonal Antibody
$286.50 - $1067.00
Specifications
Antigen | IL-22 |
---|---|
Clone | MT231 |
Concentration | 0.5 mg/ml |
Applications | ELISA |
Classification | Monoclonal |
Description
IL-22 Monoclonal antibody specifically detects IL-22 in Mouse, Rat samples. It is validated for ELISASpecifications
IL-22 | |
0.5 mg/ml | |
Monoclonal | |
Purified | |
RUO | |
Mouse, Rat | |
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL-D110, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
Recombinant mouse IL-22 | |
Primary | |
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
MT231 | |
ELISA | |
Unconjugated | |
Mouse | |
Biologically Active Proteins, Cytokine Research, Innate Immunity | |
0.2 um filtered PBS | |
50616 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title