Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-22 Rabbit anti-Human, Clone: 34, Novus Biologicals™
SDP

Catalog No. NB021783 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021783 100 μg
NB021784 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021783 Supplier Novus Biologicals Supplier No. NBP290160100UL

Rabbit Monoclonal Antibody

IL-22 Monoclonal specifically detects IL-22 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen IL-22
Applications ELISA
Classification Monoclonal
Clone 34
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL-D110, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
Host Species Rabbit
Immunogen Recombinant Human IL-22 Protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 50616
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.