Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-22 Receptor alpha Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA130723
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA130723 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA130723 Supplier Invitrogen™ Supplier No. PA130723
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2 whole cell lysate.. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

A novel cytokine, designated IL-TIF for IL-10 related T cell-derived inducible factor and IL-22, was recently identified. The receptor for IL-22 (IL-22R, also termed CRF2-9 and IL-TIF-R1 chain) is a new member of the class II cytokine receptor family. IL-22R forms a complex with IL-10 receptor beta chain and mediates IL-22 signaling. IL-22 and its receptor activate JAK-STAT signaling pathway. IL22R is expressed in normal liver and kidney and their cell lines HepG2 and TK-10. A soluble form of IL-22 receptor, also termed IL-22 binding protein (IL-22BP) and IL-22 receptor-alpha 2 (IL-22RA2), was identified very recently. IL-22BP prevents binding of IL-22 to the functional cell surface IL-22R complex and neutralizes IL-22 activity. LPS induces IL-22 expression, which indicates the role of IL-22 in inflammatory response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-22 Receptor alpha
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Gene Il22ra1
Gene Accession No. Q8N6P7
Gene Alias 9130219A07Rik; CRF2-9; Cytokine receptor class-II member 9; cytokine receptor family 2 member 9; ENSMUSG00000073746; il-22 receptor; IL-22 receptor subunit alpha-1; IL22R; IL-22R; IL22R1; IL22RA1; IL-22RA1; IL-22R-alpha-1; ILN; Interleukin; interleukin 22 receptor subunit alpha 1; interleukin 22 receptor, alpha 1; interleukin-22 receptor alpha chain; interleukin-22 receptor subunit alpha-1; RGD1559655; zcytoR11
Gene Symbols Il22ra1
Host Species Rabbit
Immunogen Peptide corresponding to aa 560-574 of human IL-22R precursor.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58985
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.