Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-22R alpha 1 Antibody (305405), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2770M
Description
IL-22R alpha 1 Monoclonal antibody specifically detects IL-22R alpha 1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
IL-22R alpha 1 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
305405 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R1, IL-22RA1, IL-22R-alpha-1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11 | |
Chinese hamster ovary cell line CHO-derived recombinant human IL-22 R alpha 1, Pro18-Thr228, Accession # Q8N6P7 | |
0.1 mL | |
Cytokine Research, Immunology, Signal Transduction | |
58985 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction