Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ IL-22RA-1, Rabbit, Unlabeled, Polyclonal,

Catalog No. 061077IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
061077IMI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 061077IMI Supplier MilliporeSigma™ Supplier No. 061077I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Interleukin-22 receptor subunit alpha-1 (UniProt Q8N6P7; also known as CRF2-9, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL-22R-alpha-1, IL-22RA1, ZcytoR11) is encoded by the IL22RA1 (also known as IL22R) gene (Gene ID 58985) in human. IL-22RA-1 is a member of the type II cytokine receptor family that complexes with IL-10RB and mediates JAK/STAT signaling. IL22 binding to heterodimer is greater than binding to IL22RA1 alone. IL-22RA-1 is expressed in colon, liver, lung, pancreas and kidney, however, no expression is observed in immune cells, such as monocytes, T-cells, and NK-cells. It is also detected in normal blood brain barrier endothelial cells as well as in multiple sclerosis lesions. Higher expression has been observed in synovial fluid cells from rheumatoid arthritis and spondyloarthropathy patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-22RA-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene IL22RA1, IL22R
Gene Accession No. Q8N6P7
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a sequence within the C-terminal region of human IL-22RA-1 cytoplasmic domain.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_067081
Test Specificity This polyclonal antibody targets a C-terminal sequence within the cytoplasmic domain of human IL-22 receptor subunit alpha-1.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.