Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-23 Antibody (207), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326067G
Description
IL-23 Monoclonal antibody specifically detects IL-23 in Mouse samples. It is validated for ELISASpecifications
IL-23 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
207 | |
ELISA | |
CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12B, IL-23, IL-23 subunit alpha, IL-23A, IL-23-A, IL-23p19, IL23P19P19, interleukin 12, p40, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-12 beta chain, interleukin-12 subunit beta, Interleukin-23 subunit p19, interleukin-six, G-CSF related factor, JKA3 induced upon T-cell activation, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2, P19, SGRF | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-23 (Uniprot#: Q9EQ14 and P43432, Met1-Ala196 and Met1-Ser335) | |
0.1 mL | |
Cancer, Cytokine Research, Innate Immunity | |
51561 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction