Learn More
Promega Corporation IL-23 Bioassay
Catalog No. PRJA2511 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Type:
Bioassay
Catalog No. Type
PRJA2511 Bioassay
1 options
Catalog No. PRJA2511 Supplier Promega Corporation Supplier No. JA2511
The simpler, more consistent IL-23 Bioassay is prequalified according to ICH guidelines and available in 96- and 384-well plate formats. No primary cell culture required. Supplied in a convenient thaw-and-use format, also available in cell propagation model (CPM) format.
  • A simpler, more consistent IL-23 bioassay.
  • Prequalified according to ICH guidelines.
  • Available in 96- and 384-well plate formats.
  • No primary cell culture required.
  • Supplied in convenient thaw-and-use format.
  • Also available in cell propagation model (CPM) format.
  • High sensitivity and specificity.
  • Consistent and reproducible results.
  • Suitable for research and clinical applications.
  • Streamlined workflow reduces hands-on time.
Specifications
Detection Method Bioluminescence
Type Bioassay
Cell Line IL-23
Druggable Target Cell-based Assay