Learn More
Invitrogen™ IL-23 p19 Monoclonal Antibody (23dcdp), eFluor™ 660, eBioscience™

Mouse Monoclonal Antibody
$496.00
Specifications
| Antigen | IL-23 p19 |
|---|---|
| Clone | 23dcdp |
| Concentration | 5 μL/Test |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
Description
Description: This 23dcdp monoclonal antibody reacts with the p19 subunit of human IL-23. This heterodimeric cytokine is composed of two disulfide-linked subunits, p40 and p19. It is closely related to IL-12, with which it shares the p40 subunit. The IL-23 receptor is also heterodimeric and shares the IL-12Rbeta1 chain with IL-12, while the IL-23R chain is unique to the IL-23 receptor complex. IL-23R signaling occurs through the Jak/STAT pathway and results in RORgammat expression, which promotes maintenance and proliferation of T helper 17 (Th17) cells. Dendritic cells and macrophages produce high levels of p40 and low levels of IL-23 in response to TLR2, TLR4, and TLR8 ligands and agonists of the beta-glucan receptor, Dectin-1. This is due to the presence of monomer and dimer forms of p40, while only a small portion associates with p19 to form IL-23. Our studies suggest donor variability, expression kinetics and type of stimulant can result in variation in IL-23 expression levels. Recent publications suggest the p19 subunit may also exist in the absence of association with p40. Applications Reported: This 23dcdp antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 23dcdp antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of cultured human dendritic cells. This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody...
IL-23 is a heterodimeric cytokine composed of the p40 subunit of IL-12 disulfide-linked with a protein p19. p19, like p35 of IL-12, is biologically inactive by itself. IL-23 interacts with IL-12Rbeta1 and an additional, novel beta2-like receptor subunit with STAT4 binding domain, termed IL-23R. IL-23 is secreted by activated mouse and human dendritic cells. Biological activities of mouse IL-23 are distinct from those of mouse IL-12. Mouse IL-23 was found not to induce significant amounts of IFN-γ. Mouse IL-23 does induce strong proliferation of memory T cells (but not naive T cells), whereas IL-12 has no effect on memory cells. Additionally, mouse IL-23 (but not IL-12) can activate mouse memory T cells to produce the proinflammatory cytokine IL-17. Human IL-23 has biological properties which are less distinct from human IL-12; human IL-23 induces proliferation of memory T cells and induces moderate levels of IFN-γ production by naive and memory T cells, as compared to IL-12.Specifications
| IL-23 p19 | |
| 5 μL/Test | |
| Flow Cytometry | |
| eFluor 660 | |
| Mouse | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Q9NPF7 | |
| 51561 | |
| Primary | |
| Affinity chromatography |
| 23dcdp | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgG2b κ | |
| Human | |
| IL23A | |
| il 23; IL 23 A; IL12B; Il-12b; Il12p40; Il-12p40; il23; IL-23; IL-23 subunit alpha; IL23A; IL-23A; IL-23-A; IL23P19; IL-23p19; ILN; Interleukin; interleukin 12B; interleukin 23 p19 subunit; interleukin 23 subunit alpha; interleukin 23, alpha subunit p19; interleukin-23 subunit alpha; Interleukin-23 subunit p19; interleukin-six, G-CSF related factor; JKA3 induced upon T-cell activation; MGC79388; P19; p40; RP23-388G23.1; SGRF; UNQ2498/PRO5798 | |
| IL23A | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.