Learn More
Invitrogen™ IL-23 p19 Monoclonal Antibody (fc23cpg), PerCP-eFluor™ 710, eBioscience™
Description
Description: This fc23cpg monoclonal antibody reacts with the p19 subunit of mouse IL-23, a heterodimeric cytokine made up of two disulfide-linked subunits, p40 and p19. It is closely related to IL-12, with which it shares the p40 subunit. IL-23 signals through a heterodimeric receptor expressed on activated T cells, macrophages, and dendritic cells. The receptor shares the IL-12Rbeta1 chain with IL-12, while the IL-23R chain is unique to the IL-23 receptor complex. Binding of IL-23 to its receptor activates Jak/STAT pathways similarly to IL-12, but the two cytokines exhibit distinct functions. Unlike IL-12, which induces T-bet and Th1 differentiation, IL-23 signaling results in RORgammat expression and supports CD4^+ T cells with the Th17 phenotype. Dendritic cells and macrophages produce IL-23 in response to TLR2, TLR4, and TLR8 ligands, as well as agonists of the beta-glucan receptor, Dectin-1. Our studies suggest donor, kinetics and type of stimulant can result in variation in IL-23 expression levels. Recent publications suggest the p19 subunit may also exist in the absence of association with p40. Applications Reported: This fc23cpg antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This fc23cpg antibody has been tested by intracellular staining and flow cytometric analysis of stimulated mouse bone marrow-derived dendritic cells using the Intracellular Fixation & Permeabilization Buffer Set (Prod...
Specifications
Specifications
| Antigen | IL-23 p19 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | fc23cpg |
| Concentration | 0.2 mg/mL |
| Conjugate | PerCP-eFluor 710 |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | IL23A |
| Gene Accession No. | Q9EQ14 |
| Gene Alias | il 23; IL 23 A; IL12B; Il-12b; Il12p40; Il-12p40; il23; IL-23; IL-23 subunit alpha; IL23A; IL-23A; IL-23-A; IL23P19; IL-23p19; ILN; Interleukin; interleukin 12B; interleukin 23 p19 subunit; interleukin 23 subunit alpha; interleukin 23, alpha subunit p19; interleukin-23 subunit alpha; Interleukin-23 subunit p19; interleukin-six, G-CSF related factor; JKA3 induced upon T-cell activation; MGC79388; P19; p40; RP23-388G23.1; SGRF; UNQ2498/PRO5798 |
| Show More |
Frequently Asked Questions (FAQs)
Our options will depend on the samples you are analyzing.
If cell viability is not critical, you can store your stained samples at 4 degrees C or on ice overnight in the dark and analyze the following day.
For samples stained with eFluor organic fluorochromes, we recommend that cells be suspended in 100 uL of Flow Cytometry Staining Buffer (Cat. No. 00-4222) and 100 uL of eBioscience IC Fixation Buffer (Cat. No. 00-8222); samples can be incubated for up to 3 days at 4 degrees C in the dark. Alternatively, the 1-step Fix/Lyse Solution (Cat. No. 00-5333) can be used. This is a great option when working with whole blood but also works for other cell types.
Yes, the eFluor Organic fluorochromes can be used for intracellular staining. The eFluor organic fluorochromes maintain bright signal and require minimal changes in compensation when fixed with eBioscience IC Fixation Buffer (Cat. No. 00-8222-49) and Permeabilization Buffer (Cat. No. 00-8333-56) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54, 00-5333-57) (as compared to live cells).
The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.
The eFluor Organic Dyes (eFluor 450, APC-eFluor 780, PerCP-eFluor 710, eFluor 710) are conventional fluorochromes. In contrast, the eVolve line of products are Quantum dots.
As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.