Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-23 p19 Monoclonal Antibody (G23-8), Functional Grade, eBioscience™, Invitrogen™

Catalog No. 16723285
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-723-285 500 μg
50-147-38 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-723-285 Supplier Invitrogen™ Supplier No. 16723285
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The G23-8 antibody reacts with the p19 subunit of mouse IL-23. The G23-8 antibody was generated from immunization with authentic, insect cell-expressed, recombit mouse IL-23 heterodimer. The G23-8 antibody can specifically neutralize IL-23 bioactivity with no effect on IL-12 p70 bioactivity. The use of a p19-specific capture antibody and a p40-specific detection antibody yields an IL-23 ELISA which is exquisitely specific for mouse IL-23. IL-12 p40 homodimer and IL-12 p70 were each run in the assay at 500 ng/mL with no interference or cross-reactivity observed. A panel of 20 unrelated cytokines was run in the IL-23 ELISA at 100 ng/mL with no cross reactivity observed; all values were at the limit of detection of the assay. For measurement of total p40 protein levels, the Mouse IL-12/23 Total p40 ELISA Ready-SET-Go! is available (88-7120). IL-23 is a heterodimeric cytokine composed of the p40 subunit of IL-12 disulfide-linked with a protein p19. p19, like p35 of IL-12, is biologically inactive by itself. IL-23 interacts with IL-12Rbeta1 and an additional, novel beta2-like receptor subunit with STAT4 binding domain, termed IL-23R. IL-23 is secreted by activated mouse and human dendritic cells. Biological activities of mouse IL-23 are distinct from those of mouse IL-12. Mouse IL-23 was found not to induce significant amounts of IFN-g.

IL-23 is a heterodimeric cytokine composed of the p40 subunit of IL-12 disulfide-linked with a protein p19. p19, like p35 of IL-12, is biologically inactive by itself. IL-23 interacts with IL-12Rbeta1 and an additional, novel beta2-like receptor subunit with STAT4 binding domain, termed IL-23R. IL-23 is secreted by activated mouse and human dendritic cells. Biological activities of mouse IL-23 are distinct from those of mouse IL-12. Mouse IL-23 was found not to induce significant amounts of IFN-γ. Mouse IL-23 does induce strong proliferation of memory T cells (but not naive T cells), whereas IL-12 has no effect on memory cells. Additionally, mouse IL-23 (but not IL-12) can activate mouse memory T cells to produce the proinflammatory cytokine IL-17. Human IL-23 has biological properties which are less distinct from human IL-12; human IL-23 induces proliferation of memory T cells and induces moderate levels of IFN-γ production by naive and memory T cells, as compared to IL-12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-23 p19
Applications ELISA, Functional Assay, Neutralization, Western Blot
Classification Monoclonal
Clone G23-8
Concentration 1 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2
Gene IL23A
Gene Accession No. Q9EQ14
Gene Alias il 23; IL 23 A; IL12B; Il-12b; Il12p40; Il-12p40; il23; IL-23; IL-23 subunit alpha; IL23A; IL-23A; IL-23-A; IL23P19; IL-23p19; ILN; Interleukin; interleukin 12B; interleukin 23 p19 subunit; interleukin 23 subunit alpha; interleukin 23, alpha subunit p19; interleukin-23 subunit alpha; Interleukin-23 subunit p19; interleukin-six, G-CSF related factor; JKA3 induced upon T-cell activation; MGC79388; P19; p40; RP23-388G23.1; SGRF; UNQ2498/PRO5798
Gene Symbols IL23A
Host Species Rat
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83430
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.