Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-23A/IL-23 P19 Antibody (1F6), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | IL-23A/IL-23 P19 |
---|---|
Clone | 1F6 |
Dilution | Western Blot 1:500-1:5000, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
IL-23A/IL-23 P19 Monoclonal antibody specifically detects IL-23A/IL-23 P19 in Human samples. It is validated for Western Blot, ELISASpecifications
IL-23A/IL-23 P19 | |
Western Blot 1:500-1:5000, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
IL-23, IL-23A, IL-23-A, IL-23p19, IL23P19P19, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-23 subunit alpha, Interleukin-23 subunit p19, JKA3 induced upon T-cell activation, MGC79388, SGRFIL-23 subunit alpha | |
A synthesized peptide derived from Human IL-23A/IL-23 P19 [UniProt Q9NPF7] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
1F6 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Cancer, Cytokine Research, Innate Immunity | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
51561 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title