Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-23A/IL-23 P19 Mouse anti-Human, Alexa Fluor 750, Clone: 727753, R&D Systems™

Catalog No. IC17161S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC17161S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC17161S100 Supplier R&D Systems Supplier No. IC17161S100UG
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-23A/IL-23 P19 Monoclonal specifically detects IL-23A/IL-23 P19 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IL-23A/IL-23 P19
Applications Flow Cytometry
Classification Monoclonal
Clone 727753
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9NPF7
Gene Alias IL23, IL-23, IL-23 p19/IL-12 p40, IL23A, IL-23A, IL-23-A, IL-23p19, IL-23p19/IL-12p40, IL23P19P19, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-23 subunit alpha, Interleukin-23 subunit p19, JKA3 induced upon T-cell activation, MGC79388, SGRF, SGRFIL-23 subunit alpha
Gene Symbols IL23A
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-23 p19 Arg20-Pro189 Accession # Q9NPF7
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51561
Test Specificity Detects human IL-23 p19 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) IL-23 heterodimer is observed, less than 10% cross-reactivity with recombinant mouse (rm) IL-23 heterodimer is observed, and no cross-reactivity with recombinant feline IL-23 p19, recombinant canine (rca) IL-23 p19, and recombinant rat IL-23 p19 is observed.. In Western blots, no cross-reactivity with rcaIL-23 p19 or rmIL-23 p19 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.