Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-23R Antibody (15N6C6), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227092AF647
Description
IL-23R Monoclonal specifically detects IL-23R in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IL-23R | |
Monoclonal | |
Alexa Fluor 647 | |
IBD17, IL-23 receptor, IL-23R, interleukin 23 receptor, interleukin-23 receptor | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
There are multiple isoforms of IL-23 receptor. Researchers are encouraged to use a tool like BLAST to determine the suitability of this antibody for their testing. | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, Immunohistochemistry (Paraffin) | |
15N6C6 | |
Western Blot, Immunohistochemistry-Paraffin | |
IL23R | |
A synthetic peptide corresponding to amino acids 288-315 of hIL-23 receptor was used as immunogen for this antibody. | |
0.1 mL | |
Immunology | |
149233 | |
Human, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction