Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-23R Antibody (753317) [DyLight 594], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB16861M
Description
IL-23R Monoclonal antibody specifically detects IL-23R in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
IL-23R | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
753317 | |
Flow Cytometry, CyTOF-ready | |
IBD17, IL-23 receptor, IL-23R, interleukin 23 receptor, interleukin-23 receptor | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-23 R, Gly24-Asp372, Accession # Q8K4B4 | |
0.1 mL | |
Immunology | |
149233 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction