Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-23R Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983306
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3306 Supplier Bioss Supplier No. BS1460R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL-23R is a 629 aa, novel subunit of the receptor for cytokine IL23A/IL23 which pairs with the receptor molecule IL12RB1/IL12R beta 1and thus take part in IL23A signaling. Signal transduction analysis confirms that this protein associates constitutively with Janus kinase 2 (JAK2), and also binds to transcription activator STAT3 in a ligand-dependent manner. Alternative splicing results in six spliced isoforms of IL-23R. Reports suggest that IL-23R plays an important role in psoriasis pathogenesis and is also a potential therapeutic target in inflammatory bowel disease.

Specifications

Antigen IL-23R
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Il23r
Gene Accession No. Q5VWK5
Gene Alias IBD17; IL 23R; IL-23 receptor; Il23r; IL-23R; interleukin 23 receptor; interleukin 23 receptor-like; Interleukin-23 receptor
Gene Symbols Il23r
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human IL-23R.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149233
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.