Learn More
IL-23R Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | IL-23R |
---|---|
Concentration | 300 μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-8558
![]() ![]() |
Proteintech Group Inc
271631AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The IL-23R antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human IL-23R. This antibody recognizes human, mouse antigen. The IL-23R antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
IL-23R | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human, Mouse | |
IgG |
300 μg/mL | |
Polyclonal | |
Rabbit | |
149233 | |
Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.