Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-24 Antibody (MM0398-6F48), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211702PE
Description
IL-24 Monoclonal antibody specifically detects IL-24 in Human samples. It is validated for Western BlotSpecifications
IL-24 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
MM0398-6F48 | |
Western Blot | |
C49A, FISP, IL10B, IL-24, interleukin 24, mda-7, MDA7MOB5, melanocyte-associated Mda-7, melanoma differentiation association protein 7, Melanoma differentiation-associated gene 7 protein, Mob-5, ST16interleukin-24, suppression of tumorigenicity 16 (melanoma differentiation), Suppression of tumorigenicity 16 protein | |
Human recombinant IL-24 | |
0.1 mL | |
Apoptosis, Cancer | |
11009 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction