Learn More
Invitrogen™ IL-25 Monoclonal Antibody (182203), APC

Description
This antibody shows 100% cross-reactivity with recombinant mouse IL-17E and no cross-reactivity with recombinant human (rh) IL-17, rhIL-17B, rhIL-17C, or rhIL-17F.
Specifications
Specifications
| Antigen | IL-25 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 182203 |
| Conjugate | APC |
| Formulation | Buffered saline solution with BSA and 0.09% sodium azide |
| Gene | IL25 |
| Gene Accession No. | Q9H293 |
| Gene Alias | D17Wsu104e; il 17e; Il17e; IL-17E; Il25; IL-25; ILN; Interleukin; interleukin 17E; interleukin 25; interleukin-17E; Interleukin-25; Ly6elg; Mydgf; myeloid derived growth factor; myeloid-derived growth factor; RGD1561632; UNQ3120/PRO10272; UPF0556 protein C19orf10 homolog |
| Gene Symbols | IL25 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.