Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-25 Monoclonal Antibody (8B1)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA516230

Catalog No. PIMA516230


Only null left
Add to Cart

Description

Description

IL-25 Monoclonal Antibody for Western Blot, Flow

Interleukin 25 (IL-25) is a member of the IL-17 family and is identified as a proinflammatory cytokine which is produced by Th17 effector cells thus enabling the Th17 subset to initiate or maintain the autoimmune process in diseases such as Rheumatoid Arthritis. IL-25 also induces activation of NF-kappa-B and stimulates production of the proinflammatory chemokine IL-8. IL-25 itself is activated through IL-23, IL-5, and TGF-beta.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-25
Monoclonal
1.0 mg/mL
PBS with 0.05% sodium azide
Q9H293
IL25
Full length recombinant IL-25.
100 μg
Primary
Human
Antibody
IgG1 κ
Flow Cytometry, Western Blot
8B1
Unconjugated
IL25
D17Wsu104e; il 17e; Il17e; IL-17E; Il25; IL-25; ILN; Interleukin; interleukin 17E; interleukin 25; interleukin-17E; Interleukin-25; Ly6elg; Mydgf; myeloid derived growth factor; myeloid-derived growth factor; RGD1561632; UNQ3120/PRO10272; UPF0556 protein C19orf10 homolog
Mouse
Protein G
RUO
64806
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.