Learn More
Invitrogen™ IL-25 Polyclonal Antibody, Biotin
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA186925
Description
Reconstitute with 0.5 mL PBS containing 0.1% BSA. Prior to reconstitution, store at 4ºC. After reconstitution, store undiluted at -20ºC, avoiding freeze/thaw cycles. Rabbit anti Mouse SF-20 antibody recognizes mouse stromal cell-derived growth factor 20 (SF-20), otherwise known as interleukin-25 (IL-25), a 166 amino acid secreted growth factor and member of the UPF0556 family.
Interleukin 25 (IL-25) is a member of the IL-17 family and is identified as a proinflammatory cytokine which is produced by Th17 effector cells thus enabling the Th17 subset to initiate or maintain the autoimmune process in diseases such as Rheumatoid Arthritis. IL-25 also induces activation of NF-kappa-B and stimulates production of the proinflammatory chemokine IL-8. IL-25 itself is activated through IL-23, IL-5, and TGF-beta.
Specifications
IL-25 | |
Polyclonal | |
PBS with no preservative; pH 7.2 | |
0 | |
IL25 | |
Recombinant mouse SF-20. | |
50 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
ELISA, Western Blot | |
Biotin | |
IL25 | |
D17Wsu104e; il 17e; Il17e; IL-17E; Il25; IL-25; ILN; Interleukin; interleukin 17E; interleukin 25; interleukin-17E; Interleukin-25; Ly6elg; Mydgf; myeloid derived growth factor; myeloid-derived growth factor; RGD1561632; UNQ3120/PRO10272; UPF0556 protein C19orf10 homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
140806 | |
-20°C | |
Lyophilized |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.