Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-26/AK155 Antibody (510414), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB13751A
Description
IL-26/AK155 Monoclonal antibody specifically detects IL-26/AK155 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
IL-26/AK155 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
510414 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
AK155IL-26interleukin-26, interleukin 26, Protein AK155 | |
E. coli-derived recombinant human IL-26/AK155, Lys22-Gln171, Accession # Q9NPH9.1 | |
0.1 mL | |
Cytokine Research | |
55801 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction