Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546973
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546973 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546973 Supplier Invitrogen™ Supplier No. PA546973
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Less than 5% cross-reactivity with recombinant human IL-10 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

IL26 is a member of the IL10 family of cytokines. It is a secreted protein and may function as a homodimer. This protein is thought to contribute to the transformed phenotype of T cells after infection by herpesvirus samimiri.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-26
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene IL26
Gene Accession No. Q9NPH9
Gene Alias AK155; il 26; IL26; IL-26; ILN; Interleukin; interleukin 26; interleukin-26; Protein AK155
Gene Symbols IL26
Host Species Goat
Immunogen E. coli-derived recombinant human IL-26 Lys22-Gln171.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55801
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.