Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-27 Antibody (355025), Alexa Fluor™ 750, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB18342S
Description
IL-27 Monoclonal antibody specifically detects IL-27 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
IL-27 | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
355025 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
IL-27, IL-27 p28 subunit, IL-27 subunit alpha, IL27A, IL-27-A, IL27-A, IL27p28, IL30, interleukin 27, interleukin 30, interleukin-27 subunit alpha, MGC71873, p28IL-27A | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-27 heterodimer, Tyr19-Pro228 (EBI3), Phe29-Ser234 (p28), Accession # O35228 (EBI3) and Q8K3I6 (p28) | |
0.1 mL | |
Cell Cycle and Replication | |
246778 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction