Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-27 p28/IL-30 Antibody (234205), PE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1834P
Description
IL-27 p28/IL-30 Monoclonal antibody specifically detects IL-27 p28/IL-30 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
IL-27 p28/IL-30 | |
Monoclonal | |
PE | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
234205 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
IL27,IL-27,IL27A,IL-27A,IL27p28,IL-27p28,IL30,p28 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse IL-27, Tyr19-Pro228 (EBI3) and Phe29-Ser234 (p28), Accession # O35228 (EBI3) and Q8K3I6 (p28) | |
0.5 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction