Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-27 p28 (IL-30) Mouse anti-Mouse, PE, Clone: MM27.7B1 (also known as MM27-7B1), BD Phrmingen™

Catalog No. BDB568409 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB568409 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB568409 Supplier BD Biosciences Supplier No. 568409
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-27 p28 (IL-30) Monoclonal antibody specifically detects IL-27 p28 (IL-30) in Mouse samples. It is validated for Intracellular staining (flow cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-27 p28 (IL-30)
Applications Flow Cytometry, Control
Classification Monoclonal
Clone MM27.7B1 (also known as MM27-7B1)
Concentration 0.2 mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Il27; IL-27 alpha; Il27a; IL27-A; IL-27-A; IL-27α; IL-27 p28; Il30; IL-30
Host Species Mouse
Immunogen Mouse IL-27
Purification Method Affinity Chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246779
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.