Learn More
IL-27/p28 Monoclonal Antibody (3H12.F10), Rockland™

Rat Monoclonal Antibody
$174.73 - $651.91
Specifications
Antigen | Mouse IL-27/p28 |
---|---|
Clone | 3H12.F10 |
Concentration | 1 mg/mL |
Applications | ELISA, Flow Cytometry, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1509
![]() ![]() |
Rockland Immunochemicals
210501B54 |
100 μg |
Each for $651.91
|
|
|||||
50-199-1510
![]() ![]() |
Rockland Immunochemicals
210501B54S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. IL-27 / p28 antibody is purified by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. This antibody is specific for mouse and rat p28 protein. Cross-reactivity with IL-27 from other sources has not been determined.
IL-27 (Interleukin 27) is a cytokine belonging to the IL-12 family, and is a heterodimeric cytokine composed of EBI3 and p28 subunits. IL-27 is produced by activated dendritic cells and macrophages in response to Toll-like receptor ligands and pro-inflammatory cytokines. IL-27 promotes CD4+ T cell differentiation to the Th1 lineage by inducing expression of the transcription factor T-bet and up regulating IL-12R beta2, which in turn suppresses Th2 and Th17 differentiation and proliferation. IL-27 is a recently discovered member of the IL-6/IL-12 family of proinflammatory and immunoregulatory cytokines. IL-27 acts by binding to its receptor WSX-1 and gp130 which results in the activation of a Jak/STAT signaling cascade, suggesting IL-27 involvement in the regulation of immune processes. It has been suggested that IL-27 can also be used as a therapeutic agent against cancer as it can also induce tumor-specific anti-tumor activity mediated through CD8+ T-cells, IFN-gamma, and T-bet. In humans, the gene is located on the p arm of chromosome 16.Specifications
Mouse IL-27/p28 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
cytokine receptor; EBI3; EBI-3; EBV-induced gene 3 protein; Epstein-Barr virus induced 3; Epstein-Barr virus induced gene 3; Epstein-Barr virus induced gene 3 (similar to Interleukin-27 beta chain precursor); Epstein-Barr virus-induced gene 3 protein; Epstein-Barr virus-induced gene 3 protein homolog; Il27; IL-27; IL27 p28; IL-27 p28 subunit; IL27 subunit; IL-27 subunit alpha; IL-27 subunit beta; Il27a; IL-27A; IL-27-A; IL27-A; IL27B; IL-27B; IL27p28; IL-27p28; IL30; IL35 subunit; ILN; Interleukin; interleukin 27; interleukin 30; Interleukin-27 subunit alpha; Interleukin-27 subunit beta; Interleukin-30; MGC71873; M-IL-27 (p28); p28; RGD1561420 | |
Ebi3, IL27 | |
IgG2a κ | |
-20°C, Avoid Freeze/Thaw Cycles | |
IL27 |
3H12.F10 | |
ELISA, Flow Cytometry, Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
O35228, Q8K3I6 | |
246779, 50498 | |
Anti-IL-27/p28 monoclonal antibody was produced in rats by repeated immunizations with mature length recombinant mouse p28 protein (produced in E.coli) followed by hybridoma development. | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.