Learn More
Invitrogen™ IL-27 p28 Recombinant Rabbit Monoclonal Antibody (7H7L10)
Description
Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Specifications
Specifications
| Antigen | IL-27 p28 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 7H7L10 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2-7.4 |
| Gene | Il27 |
| Gene Accession No. | Q8NEV9 |
| Gene Alias | IL27; IL-27; IL-27 p28 subunit; IL-27 subunit alpha; Il27a; IL-27A; IL-27-A; IL27-A; IL27p28; IL-27p28; IL30; IL-30; ILN; Interleukin; interleukin 27; interleukin 30; Interleukin-27; interleukin-27 subunit alpha; Interleukin-30; p28 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.