Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-27 Rabbit anti-Human, Clone: 1, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP28926250UL
Description
IL-27 Monoclonal specifically detects IL-27 in Human samples. It is validated for ELISA.Specifications
IL-27 | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
1 | |
ELISA 1:5000-1:10000 | |
IL-27, IL-27 p28 subunit, IL-27 subunit alpha, IL27A, IL-27-A, IL27-A, IL27p28, IL30, interleukin 27, interleukin 30, interleukin-27 subunit alpha, MGC71873, p28IL-27A | |
Recombinant Human IL-27 Protein | |
50 μg | |
Cell Cycle and Replication | |
246778 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction