Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-27R alpha/WSX-1/TCCR Antibody (34N4G11) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50 - $489.50

Specifications

Antigen IL-27R alpha/WSX-1/TCCR
Clone 34N4G11
Concentration 1.0 mg/ml
Dilution Flow Cytometry 1 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/ml, Flow (Cell Surface), CyTOF-ready
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP219015SS
SDP
View Documents
Novus Biologicals
NBP219015SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP219015
SDP
View Documents
Novus Biologicals
NBP219015
0.1 mg
Each for $489.50
Only null left
Add to Cart
 
Description

Description

IL-27R alpha/WSX-1/TCCR Monoclonal specifically detects IL-27R alpha/WSX-1/TCCR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface), CyTOF-ready.
Specifications

Specifications

IL-27R alpha/WSX-1/TCCR
1.0 mg/ml
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Unconjugated
Mouse
Apoptosis, Cytokine Research
Q6UWB1
9466
A portion of amino acids 200-250 of human IL-27R (WSX-1) was used as the immunogen for this antibody.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
34N4G11
Flow Cytometry 1 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/ml, Flow (Cell Surface), CyTOF-ready
Monoclonal
Purified
RUO
Human
class I cytokine receptor, CRL1IL-27R, Cytokine receptor-like 1, IL27R, IL-27RA, IL-27R-alpha, interleukin 27 receptor, alpha, interleukin-27 receptor subunit alpha, TCCRIL-27 receptor subunit alpha, T-cell cytokine receptor type 1, Type I T-cell cytokine receptor, WSX-1, WSX1IL-27R subunit alpha, zcytor1
IL27RA
IgG2b κ
Protein G purified
IL-27R (WSX-1) is 65-70 kDa protein and is highly expressed in lymphoid tissues such as spleen, lymph nodes and peripheral blood leukocytes including CD4+ and CD8+ T cells, monocytes, DCs and NK cells
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.