Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-27R alpha/WSX-1/TCCR Antibody (34N4G11), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP219015FR
Description
IL-27R alpha/WSX-1/TCCR Monoclonal specifically detects IL-27R alpha/WSX-1/TCCR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface).Specifications
IL-27R alpha/WSX-1/TCCR | |
Monoclonal | |
DyLight 680 | |
Q6UWB1 | |
IL27RA | |
A portion of amino acids 200-250 of human IL-27R (WSX-1) was used as the immunogen for this antibody. | |
0.1 mL | |
Apoptosis, Cytokine Research | |
9466 | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
34N4G11 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface) | |
class I cytokine receptor, CRL1IL-27R, Cytokine receptor-like 1, IL27R, IL-27RA, IL-27R-alpha, interleukin 27 receptor, alpha, interleukin-27 receptor subunit alpha, TCCRIL-27 receptor subunit alpha, T-cell cytokine receptor type 1, Type I T-cell cytokine receptor, WSX-1, WSX1IL-27R subunit alpha, zcytor1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
IL-27R (WSX-1) is 65-70 kDa protein and is highly expressed in lymphoid tissues such as spleen, lymph nodes and peripheral blood leukocytes including CD4+ and CD8+ T cells, monocytes, DCs and NK cells | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction