Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-27R alpha/WSX-1/TCCR Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP176728SS
Description
IL-27R alpha/WSX-1/TCCR Polyclonal specifically detects IL-27R alpha/WSX-1/TCCR in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
IL-27R alpha/WSX-1/TCCR | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
class I cytokine receptor, CRL1IL-27R, Cytokine receptor-like 1, IL27R, IL-27RA, IL-27R-alpha, interleukin 27 receptor, alpha, interleukin-27 receptor subunit alpha, TCCRIL-27 receptor subunit alpha, T-cell cytokine receptor type 1, Type I T-cell cytokine receptor, WSX-1, WSX1IL-27R subunit alpha, zcytor1 | |
Rabbit | |
70 kDa | |
0.025 mg | |
Primary | |
Human | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL | |
NP_004834 | |
IL27RA | |
Antibody was raised against a synthetic peptide corresponding to amino acids near the amino terminus of human TCCR precursor. The immunogen is located within the first 50 amino acids of TCCR. Amino Acid Squence: GSAGPLQCYGVGPLGD | |
Affinity Purified | |
Apoptosis, Cytokine Research | |
9466 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction