Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-28A/IFN-lambda 2 Antibody (6H9E6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP252495
Description
IL-28A/IFN-lambda 2 Monoclonal specifically detects IL-28A/IFN-lambda 2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
IL-28A/IFN-lambda 2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
interleukin-28A, IL28A, IL-28A, interferon, lambda 2 | |
Mouse | |
22.3 kDa | |
0.1 mg | |
Biologically Active Proteins | |
282616 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
6H9E6 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
IFNL2 | |
Purified recombinant fragment of human IL28A (AA: 1-200) expressed in E. Coli. | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction