Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-28A/IFN-lambda 2 Antibody (RN28) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP200455500UG
Description
IL-28A/IFN-lambda 2 Monoclonal specifically detects IL-28A/IFN-lambda 2 in Mouse samples. It is validated for ELISA.Specifications
IL28 | |
Monoclonal | |
0.5 mg/mL | |
ELISA 1:100-1:2000 | |
IFNL2 interferon, lambda 2, IL28A, IL-28A | |
Rat | |
Affinity Purified | |
RUO | |
282616 | |
Store at 4C. Do not freeze. |
ELISA | |
RN28 | |
Unconjugated | |
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
IFNL2 | |
The immunogen for this antibody is IL-28. | |
500 μg | |
Primary | |
Mouse | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction