Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-28A/IFN-lambda 2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$201.00 - $526.50
Specifications
Antigen | IL-28A/IFN-lambda 2 |
---|---|
Clone | 884113R |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB15873SP
![]() |
R&D Systems
MAB15873SP |
25 μg |
Each for $201.00
|
|
|||||
MAB15873100
![]() |
R&D Systems
MAB15873100 |
100 μg |
Each for $526.50
|
|
|||||
Description
IL-28A/IFN-lambda 2 Monoclonal specifically detects IL-28A/IFN-lambda 2 in Human samples. It is validated for ELISA.Specifications
IL-28A/IFN-lambda 2 | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
NP_742150 | |
282616 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
884113R | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
interleukin-28A, IFN-lambda 2, IL28A, IL-28A, interferon, lambda 2 | |
Mouse myeloma cell line NS0-derived recombinant human IL-28A/IFN-lambda 2 Val26-Val200 Accession # NP_742150 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-28A/IFN-lambda 2 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title