Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-28A/IL-28B Monoclonal Antibody (244716)
GREENER_CHOICE

Catalog No. PIMA523902
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523902 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523902 Supplier Invitrogen™ Supplier No. MA523902
Add to Cart
Add to Cart

Rat Monoclonal Antibody

In sandwich ELISAs, 100% cross-reactivity with recombinant mouse IL-28A/IFN-lambda 2 and no cross-reactivity with recombinant human (rh) IL-28A or rhIL-29 is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

IL28 is a cytokine distantly related to type I interferons and the IL-10 family. This gene, interleukin 28B (IL28B), and interleukin 29 (IL29) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA).
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-28A/IL-28B
Applications ELISA, Neutralization
Classification Monoclonal
Clone 244716
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Ifnl3
Gene Accession No. Q4VK74, Q8CGK6
Gene Alias Cytokine Zcyto20; cytokine Zcyto22; EG330496; IFL-1; IFN lambda2; Ifnl2; Ifnl3; IFN-lambda2; IFN-lambda-2; IFN-lambda-3; IFN-lambda-4; IL 28; IL 28 A; Il28; Il28a; IL-28A; Il28b; IL-28B; IL28C; IL-28C; ILN; INF-alpha; INF-lambda; interferon alpha; interferon lambda 2; interferon lambda 3; Interferon lambda2; interferon lambda-2; Interferon lambda-3; interferon, lambda 2; interferon, lambda 3; interferon, lambda 4; interferon-lambda 2; interferon-lambda2; Interleukin; interleukin 28; interleukin 28A; interleukin 28A (interferon, lambda 2); interleukin 28B; interleukin-28A; Interleukin-28B; interleukin-28C; ZCYTO20; ZCYTO22
Gene Symbols Ifnl2, Ifnl3
Host Species Rat
Immunogen E. coli-derived recombinant mouse IL-28B/IFN-lambda3 Asp20-Val193.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 330496, 338374
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.